Learn More
Endothelin 1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12191-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.Specifications
Endothelin 1 | |
Polyclonal | |
Unconjugated | |
EDN1 | |
EDN1, endothelin 1, ET 1 Big endothelin 1, ET1, HDLCQ7, PPET1, Preproendothelin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1906 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05305 | |
EDN1 | |
Endothelin 1 Fusion Protein Ag2847 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |