missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelin 1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Endothelin 1 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16839753
|
Proteintech
12191-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16829753
|
Proteintech
12191-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.Specifications
Endothelin 1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P05305 | |
1906 | |
Endothelin 1 Fusion Protein Ag2847 | |
Primary | |
-20°C | |
EDN1 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
EDN1, endothelin 1, ET 1 Big endothelin 1, ET1, HDLCQ7, PPET1, Preproendothelin 1 | |
EDN1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |