missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENTPD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13021-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
T-cadherin was initially identified as cadherin-type cell adhesion molecule expressed in various neuronal populations in a temporally and spatially restricted pattern during axon growth. T-cadherin is an atypical member of the cadherin family because it does not possess the typical transmembrane and cytoplasmic domains but is instead anchored to the plasma membrane by glycosylphosphatidylinositol (GPI) linkage. T-cadherin may play a role in maligt tumor development as loss of the chromosome locus containing the T-cadherin gene correlates with the development of a variety of cancers. Recently it has been shown that T-cadherin can act as a receptor for hexameric and high-molecular weight forms of adiponectin, suggesting that T-cadherin may also play a role in metabolic regulation.Specifications
ENTPD3 | |
Polyclonal | |
Unconjugated | |
ENTPD3 | |
CD39 antigen like 3, CD39L3, Ecto apyrase 3, Ecto ATP diphosphohydrolase 3, Ecto ATPase 3, Ecto ATPDase 3, ENTPD3, HB6, NTPDase 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
215446, 316077, 956 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75355 | |
Entpd3 | |
ENTPD3 Fusion Protein Ag4114 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |