missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENTPD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19597-1-AP-20UL
This item is not returnable.
View return policy
Description
Hydrolyzes preferentially nucleoside 5′-diphosphates, nucleoside 5′-triphosphates are hydrolyzed only to a minor extent. The order of activity with different substrates is UDP >> GDP = CDP = TDP, AMP, ADP, ATP and UMP are not substrates. Preferred substrates for isoform 2 are CTP, UDP, CDP, GTP and GDP, while isoform 1 utilizes UTP and TTP.Specifications
ENTPD4 | |
Polyclonal | |
Unconjugated | |
ENTPD4 | |
ENTPD4, KIAA0392, LALP70, LAP70, LYSAL1, NTPDase 4, UDPase, Uridine diphosphatase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361063, 67464, 9583 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9DBT4, Q9Y227 | |
Entpd4 | |
ENTPD4 Fusion Protein Ag5175 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ENTPD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction