missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Entry Port 23 mm for IN260, Standard Position Right Side Center/Top

Optional entry port for Memmert Incubators.

Brand:  Memmert™ IN260F3

1418.57 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 17557555

  • 1565.00 SEK / Each
Only null left
Estimated Shipment: 18-08-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Entry port with 23 mm diameter
  • Standard position right side center/top
Specifications

Specifications

IN260 Incubator
Entry Port
23 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Memmert™ Entry Port 23 mm for IN260, Standard Position Right Side Center/Top > For Use With: IN260

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.