Learn More
Memmert™ Entry Port Left Centre/Centre
For introducing connections at the side. Moisture tight and can be closed by flap and silicone stopper
Brand: Memmert™ IPP260ecoF0
1418.57 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Standard position left centre/centre
- Not for model size 260 with light module
- Not for model size 110 with light module
Specifications
Moisture tight | |
IPP260eco |
Product Content Correction
Product Title
Memmert™ Entry Port Left Centre/Centre > IPP260eco
Spot an opportunity for improvement?Share a Content Correction