Learn More
EOMES Sheep anti-Human, Polyclonal, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody has been used in 3 publications
Brand:  R&D Systems AF6166-SP
Encompass
Product Code. 15759012
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
EOMES Polyclonal specifically detects EOMES in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
EOMES Sheep anti-Human, Polyclonal, R&D Systems™