missing translation for 'onlineSavingsMsg'
Learn More

EOMES/TBR2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen EOMES/TBR2
Concentration 300 ÎĽg/mL
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886085
View Documents
Proteintech
28316-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16876085
View Documents
Proteintech
28316-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The EOMES/TBR2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EOMES/TBR2. This antibody recognizes human, mouse, rat antigen. The EOMES/TBR2 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

EOMES/TBR2
Western Blot, ELISA
Unconjugated
Human, Rat, Mouse
IgG
300 ÎĽg/mL
Polyclonal
Rabbit
8320
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
EOMES/TBR2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.