Learn More
EPAC2 Mouse anti-Human, Unconjugated, Clone: 1H4A10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67044-1-IG-20UL
This item is not returnable.
View return policy
Description
The EPAC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPAC2. This antibody recognizes human, mouse, rat antigen. The EPAC2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
EPAC2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
1H4A10 | |
Unconjugated | |
20 μL | |
11069 | |
IgG1 |
Product Content Correction
Product Title
EPAC2 Mouse anti-Human, Unconjugated, Clone: 1H4A10, Proteintech > 20 μL
Spot an opportunity for improvement?Share a Content Correction