missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPB41L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13440-1-AP-20UL
This item is not returnable.
View return policy
Description
Erythrocyte membrane protein band 4.1 (EPB41) is a multifunctional protein that mediates interactions between the erythrocyte cytoskeleton and the overlying plasma membrane.Specifications
EPB41L1 | |
Polyclonal | |
Unconjugated | |
EPB41L1 | |
4.1N, Band 4.1 like protein 1, EPB41L1, KIAA0338, Neuronal protein 4.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13821, 2036, 59317 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H4G0, Q9WTP0, Q9Z2H5 | |
Epb41l1 | |
EPB41L1 Fusion Protein Ag4250 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EPB41L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction