missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPB41L4A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26596-1-AP-20UL
This item is not returnable.
View return policy
Description
Members of the band 4.1 protein superfamily, including EPB41L4A, are thought to regulate the interaction between the cytoskeleton and plasma membrane (Ishiguro et al., 2000 ).Specifications
EPB41L4A | |
Polyclonal | |
Unconjugated | |
EPB41L4A | |
Band 4.1 like protein 4A, EPB41L4, EPB41L4A, NBL4, Protein NBL4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13824, 64097 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P52963, Q9HCS5 | |
EPB41L4A | |
EPB41L4A Fusion Protein Ag23923 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EPB41L4A Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction