Learn More
EPCAM Mouse anti-Human, Unconjugated, Clone: 2A2D5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66316-1-IG-20UL
Description
The EPCAM antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPCAM. This antibody recognizes human antigen. The EPCAM antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
EPCAM | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
2A2D5 | |
Unconjugated | |
20 μL | |
4072 | |
IgG2a |
Product Suggestions
Customers who viewed this item also viewed.