Learn More
EPHB1 Mouse anti-Human, Unconjugated, Clone: 1B7C12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67080-1-IG-150UL
 View more versions of this product
Product Code. 16879495
  • 4652.90 SEK / 150µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The EPHB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPHB1. This antibody recognizes human, mouse antigen. The EPHB1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
EPHB1
Monoclonal
1500 μg/mL
Mouse
Primary
Human, Mouse
Western Blot, ELISA
1B7C12
Unconjugated
150 μL
2047
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EPHB1 Mouse anti-Human, Unconjugated, Clone: 1B7C12, Proteintech > 150 μL