Learn More
Proteintech EPHB1 Mouse anti-Human, Unconjugated, Clone: 1B7C12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67080-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The EPHB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPHB1. This antibody recognizes human, mouse antigen. The EPHB1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
EPHB1 | |
Monoclonal | |
1500 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Western Blot, ELISA | |
1B7C12 | |
Unconjugated | |
20 μL | |
2047 | |
IgG2b |