Learn More
EPHB1 Mouse anti-Human, Unconjugated, Clone: 1B7C12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67080-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16879495
  • 3665.00 SEK / 150µL
Estimated Shipment: 31-03-2023
to see stock.
Description
Description
The EPHB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPHB1. This antibody recognizes human, mouse antigen. The EPHB1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
EPHB1
Monoclonal
1500 μg/mL
Mouse
Primary
Mouse, Human
ELISA, Western Blot
1B7C12
Unconjugated
150 μL
2047
IgG2b