Learn More
Eppendorf™ Membrane Filters for Easypet™ 3 Electronic Pipet Controller
Filters for the Easypet 3 Electronic Pipet Controller
Brand: Eppendorf™ 4421601009
1254.57 SEK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Description
Filters for the Easypet 3 Electronic Pipet Controller
- Pack of five
- For Easypet 3
- Sterile
- PTFE
Order Info
Also available: Easypet 3 Electronic Pipet Controller (Cat. Nos. 12-654-100); rechargeable lithium battery (12-654-104)
Specifications
PTFE | |
0.45 ÎĽm | |
Sterile | |
Easypet™ 3 electronic pipet controller |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
New Offer Save Now -
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
New Offer Save Up to 78%