Learn More
EPS8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12455-1-AP-150UL
 View more versions of this product
Product Code. 16880174
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This gene encodes a member of the EPS8 family. This protein contains one PH domain and one SH3 domain. It functions as part of the EGFR pathway, though its exact role has not been determined. Highly similar proteins in other organisms are involved in the transduction of signals from Ras to Rac and growth factor-mediated actin remodeling. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
Specifications
Specifications
EPS8
Polyclonal
Unconjugated
EPS8
EPS8
Rabbit
Antigen Affinity Chromatography
RUO
2059
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.49 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q12929
EPS8
EPS8 Fusion Protein Ag3121
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EPS8 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated