missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERGIC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ERGIC2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879313
|
Proteintech
11927-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869313
|
Proteintech
11927-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Possible role in transport between endoplasmic reticulum and Golgi.Specifications
ERGIC2 | |
Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q96RQ1 | |
51290 | |
ERGIC2 Fusion Protein Ag2603 | |
Primary | |
-20°C | |
ERGIC2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
cd002, CDA14, ERGIC and Golgi 2, ERGIC2, Erv41, PTX1 | |
ERGIC2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ERGIC2 Rabbit anti-Human, Polyclonal, Proteintech