missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESD Mouse anti-Human, Unconjugated, Clone: 2F9A7, Proteintech
Mouse Monoclonal Antibody
1706.43 SEK - 4863.32 SEK
Specifications
Antigen | ESD |
---|---|
Clone | 2F9A7 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
The ESD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ESD. This antibody recognizes human antigen. The ESD antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ESD | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
2098 | |
Primary |
2F9A7 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ESD Mouse anti-Human, Unconjugated, Clone: 2F9A7, Proteintech