missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14007-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The human ER-beta is a newly discovered estrogen receptor initially cloned and characterized from testis. The size and structure of ER-beta is very similar to ER-alpha with the ligand and DNA binding domains being highly conserved, while the amino terminus which serves as their transactivation domain has diverged significantly. Similar in function to ER-alpha ER-beta binds to estrogen with a high affinity and regulates estrogen specific gene activation through direct interaction with estrogen response elements (ERE′s).Specifications
ESR2 | |
Polyclonal | |
Unconjugated | |
ESR2 | |
ER BETA, Erb, ESR BETA, ESR2, ESRB, ESTRB, estrogen receptor 2 (ER beta), Estrogen receptor beta, NR3A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13983, 2100, 25149 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08537, Q62986, Q92731 | |
ESR2 | |
ESR2 Fusion Protein Ag5103 | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |