missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Ethernet Connection Cable for Computer Interface for UN75

Ethernet connection cable for use with Memmert Ovens.

Brand:  Memmert™ UN75E06189

329.11 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 17580486

  • 344.00 SEK / Each
Only null left
Estimated Shipment: 05-09-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • 5 m
  • Accessory
Specifications

Specifications

UN75 Oven
USB Connection Cable
5 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Memmert™ Ethernet Connection Cable for Computer Interface for UN75 > For Use With: UN75

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.