missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12280-1-AP-20UL
This item is not returnable.
View return policy
Description
ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes.Specifications
ETS2 | |
Polyclonal | |
Unconjugated | |
ETS2 | |
ETS2, ETS2IT1, Protein C ets 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2114, 23872 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P15036, P15037 | |
ETS2 | |
ETS2 Fusion Protein Ag2929 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ETS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction