Learn More
Heidolph Evaporating flask 20 L
Brand:  Heidolph 15-300-003-21
13981.41 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Evaporating flask 20 L
Product Code. 17382720
  • 14180.00 SEK / Each
Only null left
Estimated Shipment: 11-08-2025
Product Title
Heidolph Evaporating flask 20 L >