missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVI2B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24891-1-AP-20UL
This item is not returnable.
View return policy
Description
CD361, also known as EVI2B (Ecotropic Viral Integration site 2B) or EVDB, is a poorly characterized type I transmembrane protein, expressed from one of three genes embedded in intron 27b of the neurofibromatosis type 1 (NF1) gene. The DNA strand that is transcribed to produce CD361 is the complementary one to the strand encoding NF1. Murine homolog to human CD361 is associated with ecotropic viral insertions, which have been implicated in the expression of murine myeloid leukemias. CD361 has been also reported to be involved in melanocyte and keratinocyte differentiation. However, it is expressed mainly in peripheral blood and bone marrow.Specifications
EVI2B | |
Polyclonal | |
Unconjugated | |
EVI2B | |
CD361, D17S376, EVDB, EVI 2B, EVI2B, Protein EVI2B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2124 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P34910 | |
EVI2B | |
EVI2B Fusion Protein Ag19511 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EVI2B Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction