Learn More
Wheaton™ Extended Controlled Dropper Tip
Shop All Wheaton Science Products Products
Click to view available options
Color:
Natural
Purple
White
Yellow
Description:
Tip size: 13mm
Tip size: 20mm
Tip size: 8mm
Specifications
Specifications
Color | Natural |
Description | Tip size: 20mm |
Material | LDPE |
Drop Size | 41 ÎĽL |
Product Type | Extended Controlled Dropper Tip |
Documents
Scientific Resources
Catalogs
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
Product Content Correction
Product Title
Wheaton™ Extended Controlled Dropper Tip >
Spot an opportunity for improvement?Share a Content Correction