Learn More
FABP4 Mouse anti-Human, Unconjugated, Clone: 3E7E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67167-1-IG-150UL
 View more versions of this product
Product Code. 16879645
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The FABP4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FABP4. This antibody recognizes human, mouse, rat, pig antigen. The FABP4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
FABP4
Monoclonal
747 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry
3E7E1
Unconjugated
150 μL
2167
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FABP4 Mouse anti-Human, Unconjugated, Clone: 3E7E1, Proteintech > 150 μL