Learn More
FABP6 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | FABP6 |
---|---|
Concentration | 333 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892164
|
Proteintech
13781-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882164
|
Proteintech
13781-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FABP6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FABP6. This antibody recognizes human, mouse antigen. The FABP6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
FABP6 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Mouse | |
IgG |
333 ÎĽg/mL | |
Polyclonal | |
Rabbit | |
2172 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.