missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor X Mouse anti-Human, Clone: 3D3F11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66753-1-IG-150UL
This item is not returnable.
View return policy
Description
F10 undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR. Two chains of the factor are held together by 1 or more disulfide bonds; the light chain contains 2 EGF-like domains, while the heavy chain contains the catalytic domain which is structurally homologous to those of the other hemostatic serine proteases. The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrisic pathway), or by factor VIIa (in the extrinsic pathway). The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting. Mutations of this gene result in factor X deficiency, a hemorrhagic condition of variable severity.Specifications
Factor X | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P00742 | |
F10 | |
Factor X Fusion Protein Ag11664 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
3D3F11 | |
Unconjugated | |
F10 | |
coagulation factor X, F10, Factor X, FX, FXA, Stuart factor, Stuart Prower factor | |
Mouse | |
Protein G | |
RUO | |
2159 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Factor X Mouse anti-Human, Clone: 3D3F11, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction