Learn More
Memmert™ Factory Calibration Certificate For 3 Temperatures (37/52/70°C), For Un30Pa
Memmert™ Calibration Certificate with a single measuring point is for three temperatures.
Brand: Memmert™ UN30paD00126
This item is not returnable.
View return policy
Description
- Works calibration certificate for 3 temperatures: +37°C, +52°C, +70°C
- Standard works calibration certificate (measuring point chamber centre) at +80°C
Specifications
Measuring point chamber centre | |
Certificate for 3 temperatures | |
Standard Works Calibration Certificate | |
37°C, 52°C, 70°C |
Product Content Correction
Product Title
Memmert™ Factory Calibration Certificate For 3 Temperatures (37/52/70°C), For Un30Pa >
Spot an opportunity for improvement?Share a Content Correction