missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM124B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21313-1-AP-150UL
This item is not returnable.
View return policy
Description
FAM124B is identified as a novel component of CHD7 and CHD8 containing complex. It is a mainly nuclear localized protein with a widespread expressed. Fam124B is a novel protein possibly involved in the pathogenesis of CHARGE syndrome and neurodevelopmental disorders.Specifications
FAM124B | |
Polyclonal | |
Unconjugated | |
FAM124B | |
FAM124B, Protein FAM124B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
241128, 690912, 79843 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BLQ0, Q9H5Z6 | |
FAM124B | |
FAM124B Fusion Protein Ag15963 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FAM124B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction