missing translation for 'onlineSavingsMsg'
Learn More

FAM3C Mouse anti-Human, Clone: 2E10D5, Proteintech

Mouse Monoclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen FAM3C
Clone 2E10D5
Concentration 0.62 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16887345
View Documents
Proteintech
60282-1-IG-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16877345
View Documents
Proteintech
60282-1-IG-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FAM3C is a 227 amino acid secretory protein that belongs to a novel cytokine-like family with sequence similarity to FAM3 family. FAM3C is a signaling cytokine with a GG domain, hydrophobic leader sequence and an N-terminal signal peptide. FAM3C has a cytokine activity and shows a change in expression in pancreatic cancer-derived cells. It is ubiquitously expressed in most of the tissues. Reports suggest that FAM3C plays an important role in inner ear development and may thus be involved in cell differentiation and proliferation during inner ear embryogenesis.
Specifications

Specifications

FAM3C
0.62 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
FAM3C, GS3786, ILEI, Interleukin like EMT inducer, Protein FAM3C
FAM3C
IgG1
Protein G
Antibody
2E10D5
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Unconjugated
Mouse
Human
Q92520
10447
FAM3C Fusion Protein Ag5516
Primary
-20°C
FAM3C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
FAM3C Mouse anti-Human, Clone: 2E10D5, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.