Learn More
FAM62B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24385-1-AP-20UL
This item is not returnable.
View return policy
Description
The FAM62B antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human FAM62B. This antibody recognizes human antigen. The FAM62B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
FAM62B | |
Polyclonal | |
Unconjugated | |
20 μL | |
57488 | |
IgG |
Western Blot, ELISA | |
507 μg/mL | |
Rabbit | |
Primary | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FAM62B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 20 μL
Spot an opportunity for improvement?Share a Content Correction