missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM98C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25274-1-AP-20UL
This item is not returnable.
View return policy
Description
FAM98C has two isoforms with MW 28-30 kDa and 37-40 kDa.Specifications
FAM98C | |
Polyclonal | |
Unconjugated | |
FAM98C | |
FAM98C, Protein FAM98C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
147965 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q17RN3 | |
FAM98C | |
FAM98C Fusion Protein Ag15042 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FAM98C Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction