Learn More
FBP17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27056-1-AP-20UL
 View more versions of this product
Product Code. 16885345
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The protein encoded by this gene is a member of the formin-binding-protein family. The protein contains an N-terminal Fer/Cdc42-interacting protein 4 (CIP4) homology (FCH) domain followed by a coiled-coil domain, a proline-rich motif, a second coiled-coil domain, a Rho family protein-binding domain (RBD), and a C-terminal SH3 domain. This protein binds sorting nexin 2 (SNX2), tankyrase (TNKS), and dynamin; an interaction between this protein and formin has not been demonstrated yet in human.
Specifications
Specifications
FBP17
Polyclonal
Unconjugated
FNBP1
FBP17, FNBP1, formin binding protein 1, Formin binding protein 17, hFBP17, KIAA0554
Rabbit
Antigen Affinity Chromatography
RUO
14269, 23048
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q80TY0, Q96RU3
Fnbp1
FBP17 Fusion Protein Ag25784
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FBP17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated