missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXL15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | FBXL15 |
---|---|
Concentration | 0.26 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890475
|
Proteintech
20895-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880475
|
Proteintech
20895-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FBXL15 is probably recognizes and binds to some phosphorylated proteins and promotes their ubiquitination and degradation.Specifications
FBXL15 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q9H469 | |
79176 | |
FBXL15 Fusion Protein Ag14949 | |
Primary | |
-20°C | |
FBXL15 |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
F box only protein 37, F box/LRR repeat protein 15, Fbl15, FBXL15, FBXO37, JET, PSD | |
FBXL15 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FBXL15 Rabbit anti-Human, Polyclonal, Proteintech