missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXL18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11556-1-AP-150UL
This item is not returnable.
View return policy
Description
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex.Specifications
FBXL18 | |
Polyclonal | |
Unconjugated | |
FBXL18 | |
F box/LRR repeat protein 18, Fbl18, FBXL18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231863, 678726, 80028 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96ME1 | |
Fbxl18 | |
FBXL18 Fusion Protein Ag2112 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FBXL18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction