missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | FBXO18 |
---|---|
Concentration | 0.14 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892844
|
Proteintech
14275-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882844
|
Proteintech
14275-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
FBXO18 | |
Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q8K2I9, Q8NFZ0 | |
291293, 50755, 84893 | |
FBXO18 Fusion Protein Ag5622 | |
Primary | |
-20°C | |
FBXO18 |
0.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
F box DNA helicase 1, F box only protein 18, F box protein, helicase, 18, FBH1, Fbx18, FBXO18 | |
Fbxo18 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FBXO18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech