missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RIIB/CD32b Antibody (S07-4B5), Novus Biologicals™
Rabbit Monoclonal Antibody
4085.00 SEK - 6320.00 SEK
Specifications
Antigen | Fc gamma RIIB/CD32b |
---|---|
Clone | S07-4B5 |
Concentration | 0.3 mg/mL |
Dilution | Western Blot, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18687824
|
Novus Biologicals
NBP3-19496-50ul |
50 ÎĽg |
4085.00 SEK
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18623934
|
Novus Biologicals
NBP3-19496-100ul |
100 ÎĽg |
6320.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RIIB/CD32b Monoclonal antibody specifically detects Fc gamma RIIB/CD32b in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Fc gamma RIIB/CD32b | |
0.3 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Immunology, Phospho Specific | |
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
2213 | |
IgG | |
Affinity purified |
S07-4B5 | |
Western Blot, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
A synthetic peptide of human Fc gamma RIIB/CD32b (Uniprot # P31994) | |
Primary | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fc gamma RIIB/CD32b Antibody (S07-4B5), Novus Biologicals™