Learn More
FCGR2A / CD32a Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27138-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.Specifications
FCGR2A / CD32a | |
Polyclonal | |
Unconjugated | |
FCGR2A | |
CD32, CD32A, CDw32, Fc gamma RII a, Fc gamma RIIa, FCG2, FcGR, FCGR2, FCGR2A, FCGR2A1, FcRII a, IGFR2, IgG Fc receptor II a | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2212 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P12318 | |
FCGR2A | |
FCGR2A / CD32a Fusion Protein Ag25878 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
FCGR2A / CD32a Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction