Learn More
FES Mouse anti-Human, Unconjugated, Clone: 1E1B11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67524-1-IG-20UL
Description
The FES antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FES. This antibody recognizes human antigen. The FES antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
FES | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunofluorescence, ELISA | |
1E1B11 | |
Unconjugated | |
20 μL | |
2242 | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.