Learn More
Sperian™ Honeywell™ FFP2 ML Respirator Half Mask with Valve
Click to view available options
Model:
FFP2D
Unit Size:
Pack of 10
Description
- Folding respirator packed in a tightly sealed individual bag to prevent contamination
- Fluid-resistant outer shell to reduce risk of contamination
- Ultra-soft hypoallergenic Willtech™ nose seal with absorption properties and a tight sealing film
- Preformed and colored nose bridge to save time to perfectly adjust the mask and identify its protection level
- Large internal volume with built-in chin piece to easily communicate and breathe
- Shielded exhalation valve with an exclusive design for improved protection and better breathing comfort
- Elasticated sliding straps, highly flexible and resistant, to increase safety, comfort and ease of donning/doffing
- No latex, no silicone, no PVC
Specifications
Specifications
Disposable | Single-use |
Product Type | Filtering Half Mask for Single Use |
Size | Medium |
With Exhalation Valve | Yes |
For Use With (Application) | Textile, Mining, Construction, Iron and Steel industries, Car Body Preparation Industry, Woodworking. |
Latex Free | Yes |
Material (Faceshield) | PES knit, PU foam, PU film |
Model | FFP2D |
Nose Piece Material | Polypropylene-wrapped Galvanized wire |
Quantity | 10/Pk. |
Show More |
Documents
Scientific Resources
Brochures
Applications and solutions series: Safety edition
File size: 7.7 MB
Product Content Correction
Product Title
Sperian™ Honeywell™ FFP2 ML Respirator Half Mask with Valve >
Spot an opportunity for improvement?Share a Content Correction