missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18171-1-AP-20UL
This item is not returnable.
View return policy
Description
Plays an important role in the regulation of cell proliferation and cell differentiation. Required for normal regulation of the hair growth cycle. Functions as an inhibitor of hair elongation by promoting progression from anagen, the growth phase of the hair follicle, into catagen the apoptosis-induced regression phase.Specifications
FGF5 | |
Polyclonal | |
Unconjugated | |
FGF5 | |
FGF 5, FGF5, fibroblast growth factor 5, HBGF 5, Smag 82 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14176, 2250 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12034, P15656 | |
FGF5 | |
FGF5 Fusion Protein Ag12824 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FGF5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction