Learn More
Thermo Scientific Chemicals Fibrinogen-Binding Inhibitor Peptide
Description
Inhibits the binding of fibrinogen and fibronectin
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Purification Method | HPLC |
Common Name | Fibrinogen Îł-chain dodecapeptide |
Format | Solid |
For Use With (Application) | Inhibits the binding of fibrinogen and fibronectin |
Molecular Weight (g/mol) | 1189.30 |
Product Type | Fibrinogen-Binding Inhibitor Peptide |
Quantity | 1 mg |
Regulatory Status | RUO - research use only |
Sequence | H-His-His-Leu-Gly-Gly-Ala-Lys-Gln-Ala-Gly-Asp-Val-OH |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.