missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen gamma chain Mouse anti-Human, Clone: 2B2B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66158-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is the gamma component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia. Alternative splicing results in two transcript variants encoding different isoforms.Specifications
Fibrinogen gamma chain | |
Monoclonal | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02679 | |
FGG | |
Fibrinogen gamma chain Fusion Protein Ag8817 | |
150 ÎĽL | |
Primary | |
Human | |
Antibody | |
IgM |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2B2B8 | |
Unconjugated | |
FGG | |
FGG, fibrinogen gamma chain | |
Mouse | |
Affinity Chromatography | |
RUO | |
2266 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fibrinogen gamma chain Mouse anti-Human, Clone: 2B2B8, Proteintech > 150 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction