Learn More
ROBU VitraPOR™ Filter Crucible, 15 mL
Crucibles are marked with an individual number and allow a distinct sample.
Brand:  ROBU VitraPOR™ 20152
Additional Details : Weight : 0.23460kg
 View more versions of this product
Product Code. 16882996
  • 1370.00 SEK / Pack of 10
Estimated Shipment: 17-04-2023
to see stock.
Add to basket
Description
Description
  • For analytical filtration in laboratory
  • Suitable glass adapters and rubber sleeves are available
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass 3.3
15 mL
2