Learn More
ROBU VitraPOR™ Filter Crucible, 8 mL
Crucibles are marked with an individual number and allow a distinct sample.
Brand: ROBU VitraPOR™ 20080
Additional Details : Weight : 0.17950kg
Description
- For analytical filtration in laboratory
- Suitable glass adapters and rubber sleeves are available
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
8 mL | |
0 |