missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBU VitraPOR™ Filter Crucible, 8 mL
Crucibles are marked with an individual number and allow a distinct sample.
1540.00 SEK - 2880.00 SEK
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
16852996
|
ROBU VitraPOR™
20085 |
5 |
2880.00 SEK
Pack of 10 |
Estimated Shipment: 15-04-2024
Log in to see stock. |
|||||
16842996
|
ROBU VitraPOR™
20084 |
4 |
1790.00 SEK
Pack of 10 |
Estimated Shipment: 15-04-2024
Log in to see stock. |
|||||
16802996
|
ROBU VitraPOR™
20080 |
0 |
1540.00 SEK
Pack of 10 |
Estimated Shipment: 15-04-2024
Log in to see stock. |
|||||
16822996
|
ROBU VitraPOR™
20082 |
2 |
1540.00 SEK
Pack of 10 |
Estimated Shipment: 15-04-2024
Log in to see stock. |
|||||
Description
- For analytical filtration in laboratory
- Suitable glass adapters and rubber sleeves are available
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
8 mL |