missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Crucible, 8 mL

Crucibles are marked with an individual number and allow a distinct sample.

Brand:  ROBU VitraPOR™ 20085

Additional Details : Weight : 0.17950kg

 View more versions of this product

Product Code. 16852996

  • 2880.00 SEK / Pack of 10
Estimated Shipment: 28-05-2024
to see stock.
Add to basket

Description

Description

  • For analytical filtration in laboratory
  • Suitable glass adapters and rubber sleeves are available
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
8 mL
5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers