Learn More
BRAND™ Filter Crucibles
Borosilicate glass construction. Squat form offers stability and low risk of spillage.
4055.00 SEK - 5750.00 SEK
Product Code | Brand | Capacity (Metric) | Porosity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Porosity | Price | Quantity & Availability | ||||
15212449
|
BRAND™
458024 |
30 mL | 4 |
4055.00 SEK
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
15262449
|
BRAND™
458734 |
50 mL | 4 |
5750.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
15252449
|
BRAND™
458733 |
50 mL | 3 |
5005.00 SEK
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
Description
- Borosilicate glass construction
- Supplied in packs of 10
Specifications
Borosilicate Glass | |
Filter crucible |