missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 160 mm
The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.
2735.00 SEK - 3810.00 SEK
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Sterility | Non-sterile |
No. per Pack | 1 Ea. |
Thickness (Metric) | 9.5 ± 0.5 mm |
Diameter (Metric) Membrane | 160 mm |
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
16873216
|
ROBU VitraPOR™
61600 |
0 |
2735.00 SEK
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
16883216
|
ROBU VitraPOR™
61601 |
1 |
2735.00 SEK
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
16893216
|
ROBU VitraPOR™
61602 |
2 |
2735.00 SEK
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
16803226
|
ROBU VitraPOR™
61603 |
3 |
2735.00 SEK
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
16813226
|
ROBU VitraPOR™
61604 |
4 |
3285.00 SEK
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
16823226
|
ROBU VitraPOR™
6160F |
Fine |
3810.00 SEK
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
Description
IFNA1 Monoclonal Antibody for ELISA
- Highly resistant to most chemicals and temperatures up to 500°C
- The biplane version may be required in specific installation circumstances
Specifications
Borosilicate Glass (3.3) | |
1 Ea. | |
160 mm |
Non-sterile | |
9.5 ± 0.5 mm |